Neg-50™ Frozen Section Medium
Neg-50™ Frozen Section Medium
Actual product may vary
Neg-50™ Frozen Section Medium
Neg-50™ Frozen Section Medium
Neg-50™ Frozen Section Medium
Neg-50™ Frozen Section Medium
Fisherbrand
Neg-50™ Frozen Section Medium
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
Have Questions?
Catalog NumberQuantityColor
FKS126477564 oz. per bottle, 6/cs.Clear
Catalog number FKS12647756
Price (SGD)
-
Quantity:
4 oz. per bottle, 6/cs.
Color:
Clear
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
  • Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
ColorClear
Packaging Type6/Cs.
Quantity4 oz. per bottle, 6/cs.
Unit SizePK