Powder Funnels
Powder Funnels
Actual product may vary
Powder Funnels
Powder Funnels
Powder Funnels
Fisher Catalogue
Powder Funnels
Parallel stems minimize bridging of powder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemCapacity (Metric)Height (Metric)
FKS1264243510.5cm21mm225 mL10.3 cm
FKS119079247.5cm16mm100 mL8.4 cm
FKS119979146.5cm16mm50 mL6.6 cm
Catalog number FKS12642435
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
10.5cm
Diameter (Metric) Stem:
21mm
Capacity (Metric):
225 mL
Height (Metric):
10.3 cm
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
Specifications
Diameter (Metric)Top10.5cm
Capacity (Metric)225 mL
Diameter (Metric) Stem21mm
MaterialPolypropylene
Height (Metric)10.3 cm
AutoclavableAutoclavable
Length (Metric) Stem33mm
Unit SizePK