Non-pressurized Plastic Water Deionizers
Non-pressurized Plastic Water Deionizers
Actual product may vary
Non-pressurized Plastic Water Deionizers
Non-pressurized Plastic Water Deionizers
Fisherbrand
Non-pressurized Plastic Water Deionizers
Ideal for small laboratories but also for supplying autoclaves and washers in doctors' and dentists' practices, these small compact systems provide demineralized water.
Have Questions?
Catalog NumberFlow Rate
FKS1262277550 L/hr.
Catalog number FKS12622775
Price (SGD)
-
Flow Rate:
50 L/hr.
Ideal for small laboratories but also for supplying autoclaves and washers in doctors' and dentists' practices, Thermo Scientific™ Non-Pressurized Plastic Water Deionizers are small compact systems that provide demineralized water.
  • Economical design with optimal flow-design for 100% utilization of capacity
  • Reliable water quality and environmentally friendly as the resins have long service lives and are able to be regenerate
TKA DI 425
Excellently suited for the preparation of small volumes of water to fulfill daily requirements of up to 10 liters.
Non-pressurized plastic system consisting of:
  • Practical disposable cartridge
  • Analog conductivity meter
  • Hose set
  • Wall mount (Included)
TKA DI 750
Water flows from bottom to top for particularly economical optimal utilization - ideal for daily requirements of up to 50 liters.
  • Compact Non-pressurized plastic cartridge complete with analog conductivity meter and hose set
  • Wall mount is an additional accessory, 03.1404
Specifications of the cartridges
  • Cartridge conductivity meters are 230V, 50/60 Hz
  • Max temperature is 30°C
  • Material is polytheylene
  • Connector size is R ¾ in.
Specifications
DescriptionDI 425 non-pressurized
Flow Rate50 L/hr.
MaterialPolytheylene
Filter Size425 L
For Use With (Equipment)DI 425 pressureless – replacement (Part Number 01.0426)
Purity0.1 to 20 μS/cm
TypeNon-pressurized Plastic Water Deionizer
Height (Metric)500 mm
Diameter (Metric)100 mm
Net Weight (Metric)3 kg
Unit SizeEA