Scan 1200 Automatic Colony Counter
Scan 1200 Automatic Colony Counter
Actual product may vary
Fisherbrand
Scan 1200 Automatic Colony Counter
Scan 1200 is an HD automatic colony counter. It adapts itself to all media and counts in 1 click all colonies even the smallest ones. It ensures accuracy and excellent reproducibility.
Have Questions?
Catalog number FKS12618695
Price (SGD)
111,486.00
EA
Add to cart
Price (SGD)
111,486.00
EA
Add to cart
HD automatic colony counter & inhibition zone reader, delivered with software.
  • Counting on Petri dishes (55 - 90 mm)
  • Spiral™ plating
  • Color detection: 7 colors on the same dish
  • Inhibition zone measurement
  • Counting on PetriFilmTM, RIDATM Count, Compact DryTM, filtration membrane
Specifications
Product TypeColony Counter (Automatic)
DescriptionScan 1200 is an HD automatic colony counter. It adapts itself to all media and counts in 1 click all colonies even the smallest ones. It ensures accuracy and excellent reproducibility. Images and results are saved automatically. Scan 1200 is also an inhibition zone reader. Diameter of round Petri Dishes : dia. 55, 65 and 90mm
Cell SizeMinimum size of detected colony: 0.05 mm
ColorGray
Detection MethodCounting: automatic with manual control (Counting on surface, Spiral, circle mode plated Petri dishes, Counting on chromogenic agar, PetriFilmTM, MC-media PadsTM, Compact DryTM, Easy PlateTM, filtration membranes, Inhibition zone reader)
MagnificationHD CCD color camera,HD japanese lens, Zoom x 28, Resolution: 1.2 megapixels
MaterialStainless Steel
MemoryData export to PDF report, jpg, png and bmp images, ExcelTM recountable session
ModelScan 1200
Outlet StyleUSB
Product LineColony counter
Quantity1
Total CountNumber of antibiotic paper disks: up to 7 antibiotics on a dia. 90 mm Petri dish,Reading time: 7 inhibition zone reading between 1 to 3 seconds
Dimensions (L x W x H)28.8 x 26.3 x 37.3 cm
Height (English)14.69 in.
Length (English)11.34 in.
Width (English)10.35 in.
Weight (English)20 lb
Weight (Metric)9.4 kg
Frequency50-60 Hz
Voltage100/240 V
WattageMax power: 20 W
Certifications/Compliance21 CFR Part 11, ISO 7218 and AOAC 977.27
For Use With (Application)The Scan 1200 colony counter is used in the food, environmental, pharmaceutical, cosmetics, veterinary and public institutes research
For Use With (Equipment)Counting on round Petri Dishes of dia. 55 - 90 mm,PetriFilmTM, MC-Media PadsTM, Compact DryTM, easyPlateTM, filtration membranes, Compatible with dataLinkTM or dataLinkTMpro, traceability system
Includes1 Scan™ software, 1 15 V power cord,1 USB cable, 3 validation plates, 1 user manual
Warranty3-year warranty after warranty card recording
Unit SizeEA