Technical Buffer Solution pH 4.01, Mettler Toledo™
Product Image
Fisherbrand
Technical Buffer Solution pH 4.01, Mettler Toledo™
Calibrate pH sensors with reliable, all-around pH buffers. Mettler Toledo Technical pH Buffer Solution is available in handy bottles with no-fuss labels. Six-packs provide larger quantities for convenient, economical supplies.
Have Questions?
Catalog NumberQuantity
FKS126113066 x 250 mL
Catalog number FKS12611306
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
6 x 250 mL
Request bulk or custom format
Specifications
Boiling Point100°C
ColorRed
Certificate of AnalysisSupplied
Density1g/cm3
DescriptionTechnical buffer pH 4.01, 6 x 250mL
View more
pH electrodes must be calibrated regularly; at least once a day prior to measurements and after every cleaning, regeneration or long-term storage. The calibration adjusts slope and offset of an electrode to their true values. Since an electrode is characterized by both its slope and its offset, it is advisable to do a minimum of a two point calibration for reliable measurements and better precision. It is important is that the measurements are within the calibration range.
The internationally recognized pH scale is based on standard reference materials (SRM) selected by NIST (National Institute of Standards and Technology, USA). These buffer solutions are traceable to these primary standards. The quality inspection certificate, that is available for every bottle and sachet, guarantees stated values and traceability.