Caps for PCR Tubes
Caps for PCR Tubes
Actual product may vary
Fisherbrand
Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure TypeProduct Type
FKS12609535MaxyClear, w/writing surfaceFlat Cap8-Strip Flat PCR Tube Cap with Writing Surface
FIS14-222-265ClearDomed Cap8-Strip Domed PCR Tube Cap
Catalog number FKS12609535
Price (SGD)
76.00
PK
Add to cart
Color:
MaxyClear, w/writing surface
Closure Type:
Flat Cap
Product Type:
8-Strip Flat PCR Tube Cap with Writing Surface
Price (SGD)
76.00
PK
Add to cart
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
Closure ColorClear
ColorMaxyClear, w/writing surface
Volume (Metric)0.2 mL
Quantity1250/Cs.
Closure MaterialPolypropylene
For Use With (Equipment)8-strips PCR tubes and 96-well PCR microplates
SterilityNon-sterile
MaterialPolypropylene
Closure TypeFlat Cap
FormatStrip
Product Type8-Strip Flat PCR Tube Cap with Writing Surface
Unit SizePK