Scan 300 Automatic Colony Counter
Scan 300 Automatic Colony Counter
Actual product may vary
Fisherbrand
Scan 300 Automatic Colony Counter
Automatic colony counter with a CD camera and powerful software. Colony Counter Scan 300 provides rapid, accurate, complete, and traceable results.
Have Questions?
Catalog number FKS12608695
Price (SGD)
36,992.00
EA
Add to cart
Price (SGD)
36,992.00
EA
Add to cart
Automatic colony counter, delivered with software.
  • Counting on Petri dishes (55 - 90 mm)
  • Spiral™ plating
Specifications
Product TypeColony Counter (Automatic)
DescriptionScan 300 is an automatic colony counter that includes the essential features for Petri dish counting. It is specifically adapted to the usual media in food microbiology: PCA, MRS... Diameter of round Petri Dishes : dia. 55, dia. 65, dia. 90mm
Cell SizeMinimum size of detected colony: 0.1 mm
ColorWhite
Detection MethodCounting: automatic with manual control (Counting on pour plating, Counting on surface plating, Counting on Spiral™ plating, Counting on circle plating, Automatic separation of clustered colonies, Creation of polygonal exclusion zones)
MagnificationCMOS color camera - Zoom x 28 - M12 lens
MaterialStainless Steel
MemoryData export to PDF report, jpg, png or bmp images, ExcelTM recountable session
ModelScan 300
Outlet StyleUSB
Product LineColony counter
Quantity1
Total CountCounting time: up to 1000 colonies per second
Dimensions (L x W x H)28.7 x 27.1 x 28.9 cm
Height (English)11.38 in.
Height (Metric)29cm
Length (English)11.30 in.
Length (Metric)28.5cm
Width (English)10.67 in.
Width (Metric)26.5cm
Weight (English)18 lb
Weight (Metric)8.42 kg
Frequency50-60 Hz
Voltage100/240 V
WattageMax power: 20 W
Certifications/Compliance21 CFR Part 11, ISO 7218 and AOAC 977.27
For Use With (Application)The Scan 300 colony counter is used in the food, environmental, pharmaceutical, cosmetics, veterinary and public institutes research.
For Use With (Equipment)Counting on round Petri Dishes of dia. 55 - 90 mm (Counting on pour plating, Counting on surface plating, Counting on Spiral™ plating,Counting on circle plating)
Includes1 Scan™ software, 1 15 V power cord, 1 USB cable, 2 validation plates, 1 user manual
Warranty3-year warranty after warranty card recording
Unit SizeEA