Pyrex™ Glass Graduated Cylinder, Class A
Pyrex™ Glass Graduated Cylinder, Class A
Actual product may vary
Fisherbrand
Pyrex™ Glass Graduated Cylinder, Class A
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Graduated Cylinder, Class A is scaled with white enamel graduations for high visibility against liquids of any color. Constructed with a hexagonal base, the sturdy cylinders are available in a wide range of capacities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS1259427710 mL0.2 mL
FKS1192378950 mL1 mL
FKS11933789100 mL1 mL
FKS12093333250 mL2 mL
FKS11943789500 mL5 mL
Catalog number FKS12594277
Price (SGD)
146.00
PK
Add to cart
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (SGD)
146.00
PK
Add to cart
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Cylinders, Class A are scaled with white enamel graduations for high visibility against liquids of any color. Constructed with a hexagonal base, the sturdy cylinders are available in a wide range of capacities.
  • Features: sturdy hexagonal base, white ceramic markings, wide range of capacities
  • Class A: conformity certified to German calibration regulations, imprinted with conformity symbol “H” 
  • Batch certification: inscribed with individual serial number for traceability to national standards
  • Complies: ISO 4788
Specifications
Base TypeHexagonal
Scale ColorWhite
ShapeRound
TypeGraduated Cylinder
Tolerance±0.1 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassA
Capacity (Metric)10 mL
Height (Metric)140 mm
ScaleWhite enamel
Graduations0.2 mL
Unit SizePK