Azlon™ Lightweight Polypropylene Funnels
Azlon™ Lightweight Polypropylene Funnels
Actual product may vary
Fisher Catalogue
Azlon™ Lightweight Polypropylene Funnels
Lightweight polypropylene made from food grade plastic
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) StemHeight (Metric)
FKS125914469mm78 mm
FKS1202460810mm107 mm
FKS1250145613mm149 mm
Catalog number FKS12591446
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Stem:
9mm
Height (Metric):
78 mm
Manufactured from lightweight polypropylene. Azlon™ Lightweight Polypropylene Funnels include ribs that prevent airlocks and allow rapid filtration. Autoclavable to 121 °C.
  • Lightweight polypropylene funnels with ribs that prevent air locks, allowing rapid filtration
  • Autoclavable to 121°C
  • Made from food grade plastic
Specifications
Diameter (Metric) Stem9mm
MaterialPolypropylene
Height (Metric)78 mm