1x1 image pixel for data collection
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Outer
FKS1258910510 mL60 mm30 mm
FKS107653325 L365 mm220 mm
FKS1197256825 mL70 mm42 mm
FKS11962568100 mL105 mm64 mm
FKS1198256850 mL85 mm51 mm
FKS11902619250 mL145 mm85 mm
FKS11912619500 mL180 mm105 mm
FKS11910905150 mL118 mm74 mm
Catalog number FKS12589105
Price (SGD)
261.00
PK
Add to cart
Capacity (Metric):
10 mL
Height (Metric):
60 mm
Diameter (Metric) Outer:
30 mm
Price (SGD)
261.00
PK
Add to cart
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
  • Complies with DIN ISO 1773 
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
Specifications
Certifications/ComplianceISO 1773, DIN 12353
Capacity (Metric)10 mL
MaterialBorosilicate Glass, Borosilicate Glass
GraduationsYes
Height (Metric)60 mm
ShapeConical
Writing SurfaceYes
GraduatedYes
Diameter (Metric) Outer30 mm
Closure Size11
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link