PEEK Union
PEEK Union
Actual product may vary
Fisherbrand
PEEK Union
Connect piping of varying sizes. Cytiva Life Sciences™ PEEK Unions are an important accessory for various scientific applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnections
FKS125343175/16 in. Female to 1/16 in. Male
FKS126461851/16 in. Female to 1/16 in. Female
Catalog number FKS12534317
Price (SGD)
330.00
PK
Add to cart
Connections:
5/16 in. Female to 1/16 in. Male
Price (SGD)
330.00
PK
Add to cart
  • Packsize 5
  • When ordered separately the connectors are black.
  • Made of polyetheretherketone (PEEK)
Specifications
TypeUnion
Connections5/16 in. Female to 1/16 in. Male
MaterialPEEK
Unit SizePK