Check Order Status
Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups
Actual product may vary
Fisherbrand
Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Height (Metric)
FKS12523327
1.5 mL
22.6
FKS11303814
2 mL
24.9 mm
2 Options
Catalog number
FKS12523327
Price (SGD)
61.00
PK
Add to cart
Capacity (Metric):
1.5 mL
Height (Metric):
22.6
Price (SGD)
61.00
PK
Add to cart
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
A range of crystal clear polystyrene auto analyser cups with conical bases
For Technicon™, Beckman™, Konelab™
Supplied without caps
Graduated
Specifications
Quantity
1000 Pk.
Capacity (Metric)
1.5 mL
Color
Crystal Clear
Product Type
Sample Cup
Material
Polystyrene
Disposable
Single-use
For Use With (Equipment)
Technicon, Beckman and Konelab analyzers
Height (Metric)
22.6