Pyrex™ Borosilicate Glass Crystallizing Dishes
Pyrex™ Borosilicate Glass Crystallizing Dishes
Actual product may vary
Fisher Catalogue
Pyrex™ Borosilicate Glass Crystallizing Dishes
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS125042772000 mL190 mm90 mm
Catalog number FKS12504277
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Diameter (Metric):
190 mm
Height (Metric):
90 mm
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization.
  • Complies to DIN 12337
  • Manufactured from Pyrex™ borosilicate glass
  • Suitable for use up to 400°C
  • Chemically and thermally resistant
Specifications
Certifications/ComplianceDIN 12337
Bottom ShapeFlat
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
ShapeRound
TypeCrystallizing Dish
For Use With (Application)For use up to 400°C
Height (Metric)90 mm
Diameter (Metric)190 mm
Unit SizePK