MBL™ One Mark Class AS Glass Pipets with Certification
MBL™ One Mark Class AS Glass Pipets with Certification
Actual product may vary
Fisherbrand
MBL™ One Mark Class AS Glass Pipets with Certification
Manufactured from soda lime glass with tooled jets for extra strength and to ensure correct outflow times. MBL™ One Mark Class A S Glass Pipets with Certification include markings in permanent amber stain which are fused into the glass to ensure resistance to aggressive cleaning solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ToleranceColorAccuracy
FKS1246469120 mL±0.03 mLYellow±0.03 mL
FKS1242469110 mL±0.02 mLRed±0.02 mL
Catalog number FKS12464691
Price (SGD)
107.00
PK
Add to cart
Volume (Metric):
20 mL
Tolerance:
±0.03 mL
Color:
Yellow
Accuracy:
±0.03 mL
Price (SGD)
107.00
PK
Add to cart
Manufactured from soda lime glass with tooled jets for extra strength and to ensure correct outflow times. MBL™ One Mark Class A S Glass Pipets with Certification include markings in permanent amber stain which are fused into the glass to ensure resistance to aggressive cleaning solutions.
  • Complies with ISO 648 and DIN 12691
  • Calibrated for delivery with +5s waiting time
  • Each pipette coded with individual serial number
  • Supplied with calibration certificate which allows traceability to National Standards
  • Markings in permanent amber stain which are fused into the glass ensuring highest resistance to aggressive cleaning solutions
  • Manufactured from soda lime glass with tooled jets for extra strength and to ensure correct outflow times
Specifications
Accuracy±0.03 mL
CalibrationTo Deliver
Certifications/ComplianceISO 648, DIN 12691, calibration certificate
ClassAS
MaterialSoda Lime Glass
GraduatedYes
Graduation ColorAmber
Quantity2 Pack
Tolerance±0.03 mL
ColorYellow
Volume (Metric)20 mL
TypeBulb Pipet
Unit SizePK