Pyrex™ Labware Class AS Borosilicate Glass Buret Works Certified
Pyrex™ Labware Class AS Borosilicate Glass Buret Works Certified
Actual product may vary
Fisherbrand
Pyrex™ Labware Class AS Borosilicate Glass Buret Works Certified
Manufactured from precision-bore borosilicate glass tubing for high accuracy. Pyrex™ Labware Class AS Borosilicate Glass Buret Works Certified is individually tested and supplied with certificate showing exact volume dispensed at three points on t
Have Questions?
Catalog NumberCapacity (Metric)
FKS1246347110 mL
Catalog number FKS12463471
Price (SGD)
332.00
EA
Add to cart
Capacity (Metric):
10 mL
Price (SGD)
332.00
EA
Add to cart
Manufactured from precision-bore borosilicate glass tubing for high accuracy. Pyrex™ Labware Class AS Borosilicate Glass Buret Works Certified is individually tested and supplied with certificate showing exact volume dispensed at three points on t
  • Complies with ISO 385
  • Individually tested and supplied with certificate showing exact volume dispensed at three points on the scale
  • Individual serial number and certificate provides complete traceability to National Standards
  • Each burette has a +30s waiting time before final reading can be taken
  • Manufactured from precision-bore borosilicate glass tubing for high accuracy
  • 1.5mm bore PTFE key which requires no lubrication
  • White enamel graduations
Specifications
Capacity (Metric)10 mL
Length (Metric)820 mm
MaterialBorosilicate Glass
Graduations0.02 mL
IncludesPTFE Stopcock Key
ScaleWhite
Stopcock MaterialPTFE
Tolerance±0.02 mL
TypeBuret
ClassClass AS
Certifications/ComplianceISO 385
Unit SizeEA