Check Order Status
PTFE/Silicone Preassembled Screw Caps
Actual product may vary
Fisherbrand
PTFE/Silicone Preassembled Screw Caps
Have Questions?
Catalog Number
Color
FKS12463010
Royal Blue
Catalog number
FKS12463010
Price (SGD)
867.00
PK
Add to cart
Color:
Royal Blue
Price (SGD)
867.00
PK
Add to cart
Specifications
Color
Royal Blue
Material
PTFE, Silicone
Septum
Yes
For Use With (Application)
Designed to work in Agilent or other robotic arm autosampler
Includes
9mm septa
Quantity
1000/Pk.
Type
Screw Cap