Latex Dropper Teats
Latex Dropper Teats
Actual product may vary
Fisherbrand
Latex Dropper Teats
For easy dispensing of liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS124461801.5 to 5 mL (Assorted)
FKS124061801 mL
FKS124161801.5 mL
FKS124261802.5 mL
FKS124361803.75 mL
Catalog number FKS12446180
Price (SGD)
76.00
PK
Add to cart
Volume (Metric):
1.5 to 5 mL (Assorted)
Price (SGD)
76.00
PK
Add to cart
  • Flexible rubber bulbs create suction and dispensing capacity when used with pipettes
  • Provide intuitive manual pipeting control
  • Available in different capacities
Specifications
MaterialRubber
TypeBulb
Volume (Metric)1.5 to 5 mL (Assorted)
Packaging TypeMixed
Unit SizePK