MBL™ Borosilicate Glass Stirring Rod
MBL™ Borosilicate Glass Stirring Rod
Actual product may vary
Fisherbrand
MBL™ Borosilicate Glass Stirring Rod
Minimize scratching with these glass stirring rods. MBL™ Borosilicate Glass Stirring Rods are ideal for a wide range of stirring applications.
Have Questions?
Catalog NumberLength (Metric)
FKS12441627300 mm
Catalog number FKS12441627
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
300 mm
  • Borosilicate glass rods for general stirring applications
  • With rounded ends for extra safety and to minimize scratching
Specifications
For Use With (Application)General stirring applications
IncludesRounded ends
Length (English)11.81 in.
Diameter (Metric)6 mm
Length (Metric)300 mm
MaterialBorosilicate Glass
Quantity10/Pk.
Unit SizePK