Quickfit™ Short Neck Round Bottom Flask
Actual product may vary
Fisherbrand
Quickfit™ Short Neck Round Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Short Neck Round Bottom Flask offers corrosion-free performance and resistance to thermal shock.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)
FKS12439016500 mL163 mm
Catalog number FKS12439016
Price (SGD)
635.00
Pack
Capacity (Metric):
500 mL
Height (Metric):
163 mm
Price (SGD)
635.00
Pack
Manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock
  • With highest quality Quickfit™ ground glass socket
  • Manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock
  • Comprehensive range of 45 flasks from 5ml to 10000ml
  • Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements 
Specifications
Certifications/ComplianceBody diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (Metric)163 mm
Neck StyleShort
Socket Joint29/32
Bottom ShapeRound
Unit SizePack