Choose between three types for normal or fluorescence microscopy. Azpack™ Cargille™ Immersion Oil is supplied with a listing of optical and physical properties.
Have Questions?
Catalog Number
Type
Viscosity
Quantity
FKS12423170
Immersion Oil Type B
1250 cSt
30 mL
Catalog number FKS12423170
Price (SGD)
180.00
EA
Add to cart
Type:
Immersion Oil Type B
Viscosity:
1250 cSt
Quantity:
30 mL
Price (SGD)
180.00
EA
Add to cart
Supplied with specific optical and physical properties listing
For use with normal light or fluorescence microscopy
Specifications
TypeImmersion Oil Type B
Viscosity1250 cSt
For Use With (Application)Normal light or fluorescence microscopy