1x1 image pixel for data collection
Polyethylene Clinical Waste Sack
Polyethylene Clinical Waste Sack
Actual product may vary
Fisherbrand
Polyethylene Clinical Waste Sack
Made of polyethylene. Alpha Packaging™ Polyethylene Clinical Waste Sack provides safe storage of clinical waste.
Have Questions?
Catalog NumberColor
FKS12423070Yellow
Catalog number FKS12423070
Price (SGD)
503.00
PK
Add to cart
Color:
Yellow
Price (SGD)
503.00
PK
Add to cart
  • Polyethylene
  • For incineration
Specifications
ColorYellow
Dimensions (L x W)735 x 900 mm
For Use With (Application)Clinical for incineration.
MaterialPolyethylene
Product TypeWaste Sack
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link