Color Coded Mercury Free pH Buffer, pH 10.00, Reagecon™
Product Image
Fisherbrand
Color Coded Mercury Free pH Buffer, pH 10.00, Reagecon™
Have Questions?
Catalog NumberQuantity
FKS123987471000 mL
Catalog number FKS12398747
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
ColorBlue
pH10.00
Physical FormLiquid
Quantity1000 mL
Unit SizePK
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.