Check Order Status
Aluminum Alloy Rods
Actual product may vary
Fisherbrand
Aluminum Alloy Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Aluminum Alloy Rods are manufactured from aluminum alloy.
Have Questions?
Change view
Catalog Number
Length (Metric)
FKS12398139
750 mm
FKS12378139
500 mm
2 Options
Catalog number
FKS12398139
Price (SGD)
89.00
PK
Add to cart
Length (Metric):
750 mm
Price (SGD)
89.00
PK
Add to cart
Threaded
Thread diameter 10mm
Length of rod does not include the thread
Specifications
Material
Aluminum Alloy
Length (Metric)
750 mm
Product Type
Aluminum Rods
Thread Size
10 mm
Quantity
2 Pk.