Glass Rectangular Coverslips
Glass Rectangular Coverslips
Actual product may vary
Fisherbrand
Glass Rectangular Coverslips
Glass Rectangular Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)
FKS12393128#1 (0.13 to 0.16 mm)24 mm32 mm
FKS12353128#1 (0.13 to 0.16 mm)22 mm32 mm
FKS12363128#1 (0.13 to 0.16 mm)22 mm40 mm
FKS12373128#1 (0.13 to 0.16 mm)22 mm50 mm
FKS12383138#1.5 (0.16 to 0.19 mm)22 mm50 mm
Catalog number FKS12393128
Price (SGD)
10.00
PK
Add to cart
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Length (Metric):
24 mm
Width (Metric):
32 mm
Price (SGD)
10.00
PK
Add to cart
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product TypeCoverslip
ShapeRectangle
MaterialSoda Lime Glass
Length (Metric)24 mm
Width (Metric)32 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
Quantity100 Pk.
Unit SizePK