Tyvek 600 Plus White Hooded Coverall
Tyvek 600 Plus  White Hooded Coverall
Actual product may vary
Fisherbrand
Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12392239X-Large
FKS12372239Medium
FKS12382239Large
FKS123022492X-Large
Catalog number FKS12392239
Price (SGD)
-
Size:
X-Large
  • Self-adhesive chin strap for tight seal of suit to face mask
  • Stitched and over-taped seams for protection and strength
  • Tyvek zipper and zipper flap for enhanced protection
  • Elasticated face, wrists and ankles as well as glued-in elastic waist
  • Elasticated thumb loops prevent sleeves from riding up
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as medical applications and when exposed to biological hazards
Specifications
Product TypeHooded Coveralls
SizeX-Large
MaterialHDPE
ColorWhite
Waist StyleElastic
Ankle StyleElastic
Cuff StyleThumb Loop
Neck StyleHooded
Hood TypeElastic
Closure TypeZipper with Self-adhesive Flap and Chin Flap
Seam TypeStitched, Taped
Protection ClassCategory III, Type 4-B, 5-B and 6-B
ResistanceChemicals, Liquid
Certifications/ComplianceCategory III, Type 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), (EN 1149-5)
For Use With (Application)Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing or in Research and Biosecurity Laboratories
Unit SizePK