Check Order Status
Drummond Microcaps Borosilicate Glass Capillary Tubes
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Drummond Microcaps Borosilicate Glass Capillary Tubes
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Have Questions?
Contact Us
Catalog Number
Capacity (Metric)
LengthMetric
FKS12389389
1 μL
Catalog number
FKS12389389
Price (SGD)
288.00
PK
Add to cart
Capacity (Metric):
1 μL
Price (SGD)
288.00
PK
Add to cart
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Specifications
Capacity (Metric)
1 μL
Quantity
100/Pk.
Type
Capillary Tube
For Use With (Equipment)
For TLC Sample Applicators
Includes
1 glass bulb holder assembly
Unit Size
PK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link