Amber Glass Narrow Neck Winchester Bottles with Black Urea PVDC-lined Caps
Amber Glass Narrow Neck Winchester Bottles with Black Urea PVDC-lined Caps
Actual product may vary
Fisherbrand
Amber Glass Narrow Neck Winchester Bottles with Black Urea PVDC-lined Caps
Constructed of soda lime glass. Azpack™ Amber Glass Narrow Neck Winchester Bottles with Black Urea PVDC-lined Caps contains wood pulp faced steran liner.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FKS123771081000 mL31/R6
Catalog number FKS12377108
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Closure Size:
31/R6
  • Amber soda lime glass
  • Black plastic screw cap includes wood pulp faced steran liner (PVDC)
Specifications
Capacity (Metric)1000 mL
ColorAmber
MaterialSoda Lime Glass
Quantity10/PK
Width (Metric)98mm
Closure ColorBlack
Closure IncludedYes
MouthNarrow
Height (Metric)242.6 mm
Closure LinerWood Pulp-faced PVDC
Closure MaterialUrea
Closure Size31/R6
Closure TypeScrew Cap
Unit SizePK