Presept™ Disinfectant Tablets 2.5g
Actual product may vary
Fisherbrand
Presept™ Disinfectant Tablets 2.5g
Rapid-acting disinfectant tablets.
Have Questions?
Catalog NumberVolume (Metric)
FKS123686672.5 g
Catalog number FKS12368667
Price (SGD)
-
Volume (Metric):
2.5 g
The solution is rapid acting, and is effective against bacteria, spores, fungi, algae, protozoa and viruses, (including AIDS virus) and is resistant to inactivation by organic soilage.
These 2.5g tablets contain sodium dichloroisocyanurate (NaDCC) in an effervescent base. The tablets dissolve in water to provide disinfectant solutions of predetermined concentrations, without the need to weigh or measure the active ingredient. The solution is compatible with anionic and non-ionic detergents. One tablet dissolved in 10L of water provides 140ppm available chlorine. Dilution instructions are provided in each pack. Supplied in resealable plastic drum containing 100 tablets.
Use biocides safely. Always read the label and product information before use.
Specifications
Product TypeDisinfectant Tablet
FormatTablet
Quantity100/Pk.
Volume (Metric)2.5 g
Unit SizeEach