1x1 image pixel for data collection
Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap
Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap
Actual product may vary
Fisherbrand
Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap
Allows dispensing of wash bottle in an upright or inverted position. Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.
Have Questions?
Catalog NumberCapacity (Metric)Chemical LabelClosure Color
FKS12368128250 mLIsopropanolYellow
Catalog number FKS12368128
Price (SGD)
219.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
250 mL
Chemical Label:
Isopropanol
Closure Color:
Yellow
Price (SGD)
219.00
PK
Add to cart
  • Patented valve eliminates drips and allows bottle to be used upright or inverted
  • Full color NFPA diamond
  • Colored closures for enhanced identification
  • LDPE bottles with polypropylene caps
  • Sodium hypochlorite bottles are white for light protection
  • Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.
Specifications
Capacity (Metric)250 mL
ColorTranslucent
Diameter (Metric) Outer Neck53mm
IncludesYellow Polypropylene Cap
MaterialPolyethylene
Quantity4Pack
Closure ColorYellow
Certifications/ComplianceCompliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
MouthWide
Product TypeWash Bottle
Chemical LabelIsopropanol
Closure MaterialPolypropylene
Closure TypeSafety Vented Cap
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link