Check Order Status
Transfer Pipets, PE
Actual product may vary
Fisherbrand
Transfer Pipets, PE
Ideal for small-volume liquid transfers. Gosselin™ Transfer Pipets, PE are constructed of polyethylene and graduated for convenient measuring.
Have Questions?
Catalog number
FKS12367447
Price (SGD)
635.00
PK
Add to cart
Price (SGD)
635.00
PK
Add to cart
Polyethylene construction
Graduated
Specifications
Sterility
Non-sterile
For Use With (Application)
Used in dairy, environment, industries, microbiology
Quantity
5000/Cs.
Packaging Type
500/Box
Color
Natural
Volume (Metric)
3 mL
Length (Metric)
150 mm