Stainless Steel Desktop Counter
Stainless Steel Desktop Counter
Actual product may vary
Fisher Catalogue
Stainless Steel Desktop Counter
A stainless steel tally counter for maintaining a count of objects in a group of events in sequence.
Have Questions?
Catalog NumberHeight (Metric)
FKS1236647755mm
Catalog number FKS12366477
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Height (Metric):
55mm
  • Quality 4-digit metal body mechanical counter with push button actuator and reset button
  • Ergonomically designed, this counter features a finer ring and sturdy chrome plated steel construction
  • White, 4mm high figures on a black background for easy reading
  • Counts from 0 to 9999
  • Reset by turning knob located on right-hand side in clockwise direction
  • Fits snugly into the palm of your hand
  • Desk tally counter stands on a base, which can be securely fixed using the three screw holes, on a desk or table
Specifications
Product TypeTally Counter
Displayed Text0-9999, white figures on black background
Quantity1/pack
Height (Metric)55mm
Width (Metric)55mm
For Use With (Application)Maintaining a count of objects in a group of events in sequence
IncludesStand for desk or table
Unit SizeEA