81-Well Cryogenic Storage Racks with Lid
81-Well Cryogenic Storage Racks with Lid
Actual product may vary
Fisherbrand
81-Well Cryogenic Storage Racks with Lid
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS12358192Fluorescent Pink
FKS10461411Fluorescent Green
Catalog number FKS12358192
Price (SGD)
69.00
PK
Add to cart
Color:
Fluorescent Pink
Price (SGD)
69.00
PK
Add to cart
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
Specifications
ColorFluorescent Pink
Holds81 (1.5 mL or 2.0 mL Tubes)
MaterialPolypropylene
Type81-Well Cryogenic Storage Racks with Lid
Unit SizePK