Chemware™ Cylinder
Chemware™ Cylinder
Actual product may vary
Fisherbrand
Chemware™ Cylinder
Graduated Chemware™ Cylinder made from chemically inert fluoropolymers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1234891510 mL
FKS1054525450 mL
Catalog number FKS12348915
Price (SGD)
2,583.00
EA
Add to cart
Capacity (Metric):
10 mL
Price (SGD)
2,583.00
EA
Add to cart
  • These graduated cylinders offer all the advantages of non-stick inert fluoropolymers.
  • PFA is temperature resistant from -320°F (-196°C) to +500°F (+260°C) and has the lowest metal ion content.
  • As a result, Chemware™ cylinders are excellent for trace metal analysis.
Specifications
Capacity (Metric)10 mL
MaterialPFA
Diameter (English)3.02 in.
Diameter (Metric)7.6 cm
Graduations0.5 mL
AutoclavableAutoclavable
Capacity (English)0.34 oz.
Subdivision0.5 mL
Diameter (Metric) Inside13 mm
SterilityNon-sterile
Base TypeWide
Height (English)4.6 in.
Height (Metric)118.5 mm
TypeGraduated Cylinder
Unit SizeEA