Unglazed Porcelain Pestle
Unglazed Porcelain Pestle
Actual product may vary
Fisher Catalogue
Unglazed Porcelain Pestle
Use these pestles for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Pestles are constructed from optimized laboratory porcelain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS12343338175mm
FKS10656405115mm
Catalog number FKS12343338
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
175mm
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
FinishUnglazed
Product TypePestle
MaterialPorcelain
Length (Metric)175mm
Unit SizeEA