Glazed Porcelain Mortar
Glazed Porcelain Mortar
Actual product may vary
Fisherbrand
Glazed Porcelain Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Mortars are constructed from optimized laboratory porcelain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12343328250 mL
FKS1230332870 mL
Catalog number FKS12343328
Price (SGD)
169.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
169.00
EA
Add to cart
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Capacity (Metric)250 mL
FinishUnglazed
Diameter (Metric) Inner115mm
Product TypeMortar
MaterialPorcelain
Height (Metric)63mm
Unit SizeEA