Model 120 Sonic Dismembrator
Model 120 Sonic Dismembrator
Actual product may vary
Fisherbrand
Model 120 Sonic Dismembrator
Programable system ideal for small volume liquid processing. There are 3 probe size options available which allow for processing 0.2-50 mL volumes.
Have Questions?
Catalog NumberElectrical RequirementsIncludes
FKS12337338220/230 V, 50/60 HzGenerator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Catalog number FKS12337338
Price (SGD)
13,267.00
EA
Add to cart
Electrical Requirements:
220/230 V, 50/60 Hz
Includes:
Generator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Price (SGD)
13,267.00
EA
Add to cart
Use this sonic dismembrator for effective analysis and sample preparation of small sample sizes. Fisher Scientific™ Model 120 Sonic Dismembrator offers both manual and programmable operation for consistent and reproducible results. Processes 0.2 to 50mL.
  • Processes 0.2 to 50mL
  • 120 watts
  • Programmable from 1 second to 10 hours
  • Overload protection
  • Pulse mode (1 second to 59 seconds)
  • Energy Display(Watts and Joules)
  • Available accessories include: 3 Probe sizes for individual samples, Eight tip horn for increased sample throughput, Cup Horn for indirect sonication
  • Lightweight converter for handheld operation
Specifications
Voltage220/230 V
Electrical Requirements220/230 V, 50/60 Hz
Hertz20 kHz (Ultrasonic Frequency)
Wattage120 W
Volume Range (Metric)0.2 to 50 mL
Dimensions (L x W x H)13.5 x 8 x 6 in. (34.29 x 20.32 x 15.24 cm)
Height (English)6 in.
Height (Metric)15.24 cm
Length (English)13.5 in.
Length (Metric)34.29 cm
Width (English)8 in.
Width (Metric)20.32 cm
For Use With (Application)Cell disruption, protein extraction, DNA shearing/ChIP assay
IncludesGenerator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
ModelFB120
Timer1 sec. to 10 hrs.
Unit SizeEA