Secador™ Auto Desiccator Cabinet, Vertical Model
Secador™ Auto Desiccator Cabinet, Vertical Model
Actual product may vary
Fisherbrand
Secador™ Auto Desiccator Cabinet, Vertical Model
A UV light blocking, dry environment with automatic electronic dehumidification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorCabinet StyleVoltageVolume (English)
FKS12319005ClearVertical230 V1.55 cu. ft.
FIS08-647-52ClearVertical120 V1.87 cu. ft.
FIS08-647-668ClearVertical230 V1.55 cu. ft.
FIS08-647-859AmberVertical (Tinted)230 V1.87 cu. ft.
FKS12309005ClearVertical230 V1.16 cu. ft.
Catalog number FKS12319005
Price (SGD)
5,413.00
EA
Add to cart
Color:
Clear
Cabinet Style:
Vertical
Voltage:
230 V
Volume (English):
1.55 cu. ft.
Price (SGD)
5,413.00
EA
Add to cart
A UV light blocking, dry environment with automatic electronic dehumidification.
  • Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
  • 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H
Specifications
AutoclavableNot Autoclavable
ColorClear
Cabinet StyleVertical
MaterialCopolyester
Product TypeDesiccator
Door StyleSide-swinging
For Use With (Application)Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
IncludesCarrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model3.0
No. of Shelves3
ResistanceChemicals, stains and fine cracks
Shelf TypeRemovable Perforated
Temperature Range (English)41°F to 104°F
Temperature Range (Metric)5°C to 40°C
Volume (English)1.55 cu. ft.
Dimensions (L x W)16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English)16.3 in.
Length (Metric)41.4 cm
Width (English)13.4 in.
Width (English) Door8.9 in.
Width (Metric)34.1 cm
Width (Metric) Door22.5 cm
Height (English)16.4 in.
Height (English) Opening12.3 in.
Height (Metric)41.7 cm
Depth (English)16.3 in.
Depth (Metric)41.4 cm