MiniSpin™/MiniSpin™ plus Centrifuge
MiniSpin™/MiniSpin™ plus Centrifuge
Actual product may vary
Fisherbrand
MiniSpin™/MiniSpin™ plus Centrifuge
With Eppendorf MiniSpin and MiniSpin plus centrifuges, there’s no need to share! These powerful, entry-level centrifuges are small enough that each workstation can have one. Key parts are made of metal for maximum safety, and the MiniSpin plus is fast enough for molecular biology separations.
Have Questions?
Catalog NumberElectrical RequirementsModelCapacity
FKS12313745230 V, 50/60 Hz (GB)Centrifuge MiniSpin plus (EU-IVD)12 x 1.5/2 mL
Catalog number FKS12313745
Price (SGD)
9,117.00
EA
Add to cart
Electrical Requirements:
230 V, 50/60 Hz (GB)
Model:
Centrifuge MiniSpin plus (EU-IVD)
Capacity:
12 x 1.5/2 mL
Price (SGD)
9,117.00
EA
Add to cart
  • Rotor capacity: 12 × 1.5/2.0 mL vessels, 2 × 8-tube PCR strips
  • Max. speed: 14,100 × g (14,500 rpm)
  • Extremely compact footprint
  • Acceleration and deceleration time < 13 s
  • User-friendly digital display for time and speed
  • Metal rotor bowl
  • Separate short-spin key for fast and comfortable centrifugation
  • Optimal ventilation flow reduces heating and protects temperature-sensitive samples
  • Soft-touch lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
  • Specifications
    Capacity12 x 1.5/2 mL
    Max. Speed14,500 rpm
    Max. RCF14,100 x g
    DisplayLarge LCD
    Height (English) Exterior5.12 in.
    Height (Metric) Exterior13.0 cm
    Noise Level<52 dB(A)
    Timer15 sec. to 99 min., with continuous run function
    Width (English) Exterior8.86 in.
    Width (Metric) Exterior22.5 cm
    Certifications/ComplianceUL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    IncludesWith Rotor F-45-12-11
    Safety FeaturesLid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids
    VentilatedYes
    Electrical Requirements230 V, 50/60 Hz (GB)
    Controller TypeKeypad
    Depth (Metric)239 mm
    ModelCentrifuge MiniSpin plus (EU-IVD)
    Wattage85 W
    Maximum Speed Range800 to 14,500 rpm
    Rotor IncludedYes
    RefrigeratedNo
    Profile (Acceleration/Braking)13 sec.
    Drive SystemFixed angle
    Run Time15 sec. to 30 min.
    StandardsEN ISO 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-1, EN ISO 18113-3, EN ISO 15223-1, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020