Platinum Electrodes for Hoffman Voltameter Apparatus
Actual product may vary
Platinum Electrodes for Hoffman Voltameter Apparatus
Platinum Electrodes for Hoffman Voltameter Apparatus
Fisherbrand
Platinum Electrodes for Hoffman Voltameter Apparatus
These Platinum Electrodes are mounted in rubber bungs for easy insertion into the Hoffman Voltameter Apparatus.
Have Questions?
Catalog NumberLength (Metric)
FKS12311439110 mm
Catalog number FKS12311439
Price (SGD)
502.00
Pack
Length (Metric):
110 mm
Price (SGD)
502.00
Pack
  • Pair of electrodes for use with Hoffman Voltameter Apparatus to demonstrate electrolysis of water into oxygen and hydrogen
  • Corrugated, rectangular platinum foil for increased surface area
  • Connections protected in borosilicate glass tube
  • Brass screw terminals for probes up to 4 mm diameter
  • Safe storage in glass sleeves supplied
Specifications
JunctionPlatinum
MaterialPlatinum
Temperature (Metric) OperatingAmbient
TypePlatinum Electrodes
For Use With (Equipment)Glass Hoffman Voltameter Apparatus
Length (Metric)110 mm
Unit SizePack