Alumina Combustion Boat
Alumina Combustion Boat
Actual product may vary
Fisherbrand
Alumina Combustion Boat
Aluminum oxide boats of great hardness, strength and chemical resistance.
Have Questions?
Catalog NumberQuantity
FKS123070181 Ea.
Catalog number FKS12307018
Price (SGD)
193.00
EA
Add to cart
Quantity:
1 Ea.
Price (SGD)
193.00
EA
Add to cart
  • Suitable for continuous use up to 1700° to 1750°C
  • Manufactured from Alsint 99.7 oxide ceramic
Specifications
TypeCombustion Boat
ColorWhite
Height (Metric)6 mm
Length (Metric)52 mm
MaterialAluminum Oxide
Quantity1 Ea.
Width (Metric)7.5 mm
Unit SizeEA