Check Order Status
Ultra Disposable Microtome Blades
Actual product may vary
Fisherbrand
Ultra Disposable Microtome Blades
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
Have Questions?
Change view
Catalog Number
Profile
FKS12191830
Low
FIS31-537-35
High
2 Options
Catalog number
FKS12191830
Price (SGD)
415.00
PK
Add to cart
Profile:
Low
Price (SGD)
415.00
PK
Add to cart
High- and low-profile universal blades can be used with any microtome or cryostat on the market
Unique angle profile that facilitates faster ribboning
Increased Longevity: Produces an edge that is twice as strong as traditional blades
Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
Profile
Low
For Use With (Equipment)
Microtomes, Cryostats
Disposable
Single-use
Cutting Angle
34°