Pyrex™ Labware Class B Borosilicate Glass Burette with PTFE Key
Pyrex™ Labware Class B Borosilicate Glass Burette with PTFE Key
Actual product may vary
Fisherbrand
Pyrex™ Labware Class B Borosilicate Glass Burette with PTFE Key
Manufactured from chemically resistant borosilicate glass and complies with ISO 385 standards. Pyrex™ Labware Class B Borosilicate Glass Burette with PTFE Key includes white enamel graduations for good visibility with dark colored solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1218422250 mL
FKS1213422210 mL
FKS1216422225 mL
Catalog number FKS12184222
Price (SGD)
128.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
128.00
EA
Add to cart
  • Complies with ISO 385
  • Fitted with interchangeable PTFE keys for grease-free performance, with 1.5mm bore
  • White enamel graduations for good visibility with dark colored solutions
  • Manufactured from borosilicate glass for increased chemical resistance
Specifications
Capacity (Metric)50 mL
Length (Metric)820 mm
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesPTFE Key Stopcock
ScaleWhite
Stopcock MaterialPTFE
Tolerance±0.1 mL
TypeBuret
ClassClass B
Certifications/ComplianceISO 385
Unit SizeEA