Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring
Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS120946375000 mL
FKS1197261950 mL
FKS11982619100 mL
FKS11992619250 mL
FKS11902629500 mL
FKS119126291000 mL
FKS119226292000 mL
FKS1209471710,000 mL
Catalog number FKS12094637
Price (SGD)
718.00
Each
Capacity (Metric):
5000 mL
Price (SGD)
718.00
Each
Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.
  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C
Specifications
Capacity (Metric)5000 mL
Capacity (English)1.3 gal.
ColorClear
Diameter (English)7.1 in.
For Use With (Application)Ideal for mixing and storage of culture medium
IncludesPolypropylene screw cap and pouring ring
MaterialBorosilicate Glass
No. per Pack1/PK
Quantity1 Ea.
Thread Size (Metric)GL 45
Certifications/ComplianceISO 4796
Closure IncludedYes
DisposableReusable
GraduatedYes
MouthWide
ShapeRound
Height (Metric)330 mm
Product TypeReagent Bottle
Closure MaterialPolypropylene
Closure Size45 mm
Diameter (Metric)181 mm
Closure TypeScrew Cap