Borosilicate Glass Medium Wall Rimless Test Tube
Borosilicate Glass Medium Wall Rimless Test Tube
Actual product may vary
Fisherbrand
Borosilicate Glass Medium Wall Rimless Test Tube
Manufactured from borosilicate glass with a medium wall design. Fisherbrand™ Borosilicate Glass Medium Wall Rimless Test Tubes are ideal for chemical and biological work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
FKS1208809916 mm150 mm
FKS1195220816 mm125 mm
FKS1197220818 mm150 mm
Catalog number FKS12088099
Price (SGD)
-
Diameter (Metric):
16 mm
Length (Metric):
150 mm
  • Features: rimless medium walls
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Length (Metric)150 mm
MaterialBorosilicate Glass
Quantity100 Pack
RimmedNo
Diameter (Metric)16 mm
Unit SizePK