Azlon™ PMP Cylinder with Printed Graduations
Azlon™ PMP Cylinder with Printed Graduations
Actual product may vary
Fisherbrand
Azlon™ PMP Cylinder with Printed Graduations
Printed with pin sharp blue graduations for enhanced visibility. Azlon™ PMP Cylinders with Printed Graduations provide a reliable solution for measuring the volume of liquids in a laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS12074598100 mL1 mL
FKS10190960500 mL5 mL
Catalog number FKS12074598
Price (SGD)
106.00
PK
Add to cart
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (SGD)
106.00
PK
Add to cart
Printed with pin sharp blue graduations for enhanced visibility. Azlon™ PMP Cylinders with Printed Graduations provide a reliable solution for measuring the volume of liquids in a laboratory.
  • Graduations: printed enhanced pin sharp blue 
  • Base: stable hexagonal bases with non-slip anti-suction feature
  • Calibrated: to ISO 6706 and DIN 12681
  • Class A: tolerances, conformity certified, supplied with certificate
Specifications
Capacity (Metric)100 mL
MaterialPMP
Tolerance±1 mL
Certifications/ComplianceDIN 12681, ISO 6706
ClassA
ScaleBlue
Diameter (Metric)34 mm
IncludesConformity Certificate
Quantity2PK
Graduations1 mL
AutoclavableAutoclavable
Scale ColorBlue
Base TypeHexagonal
Height (Metric)250 mm
TypeUngraduated Cylinder
Unit SizePK