Micro-Ninety™ Cleaning Solution
Micro-Ninety™ Cleaning Solution
Actual product may vary
Fisherbrand
Micro-Ninety™ Cleaning Solution
Use this concentrated cleaning solution for critical applications. Cole-Parmer™ Micro-Ninety™ Cleaning Solution cleans glass, most metals, plastic, rubber, fabric and is safe for all ultrasonic cleaners.
Have Questions?
Catalog NumberVolume (Metric)
FKS120616991 L
Catalog number FKS12061699
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1 L
  • A mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications
  • Radioactive contaminants will not reduce cleaning capability
  • Removes most biological materials
  • Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water
  • Solution is environmentally friendly
  • Contains no phosphates, CFCs, ODCs or borates
  • It safely replaces chromic acid to remove protein from glassware
  • Note: Do not use on zinc, magnesium or aluminum 
Specifications
FormLiquid
pH9.5
AppearanceYellow
Boiling Point100°C
Container TypeBottle
Volume (Metric)1 L
Unit SizeEA