Azlon™ Polypropylene Powder Funnels
Azlon™ Polypropylene Powder Funnels
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Powder Funnels
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
Have Questions?
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemHeight (Metric)
FKS12054638100mm26mm90 mm
Catalog number FKS12054638
Price (SGD)
92.00
PK
Add to cart
Diameter (Metric)Top:
100mm
Diameter (Metric) Stem:
26mm
Height (Metric):
90 mm
Price (SGD)
92.00
PK
Add to cart
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
  • Lightweight polypropylene powder funnel
  • Unbreakable in normal use
  • Parallel stem minimises 'bridging' of powder to improve flow
  • The two largest sizes have vertical sided tops
Specifications
Diameter (Metric)Top100mm
Diameter (Metric) Stem26mm
MaterialPolypropylene
Height (Metric)90 mm