Check Order Status
Pyrex™ Wide-Neck Erlenmeyer Flask
Actual product may vary
Fisherbrand
Pyrex™ Wide-Neck Erlenmeyer Flask
Manufactured from Pyrex™ borosilicate glass with white enameled graduations.
Have Questions?
Catalog Number
Capacity (Metric)
Height (Metric)
Diameter (Metric) Outer
FKS12039012
250 mL
140 mm
85 mm
Catalog number
FKS12039012
Price (SGD)
298.00
PK
Add to cart
Capacity (Metric):
250 mL
Height (Metric):
140 mm
Diameter (Metric) Outer:
85 mm
Price (SGD)
298.00
PK
Add to cart
Manufactured from PyrexTM borosilicate glass with white enameled graduations.
Complies with DIN ISO 24450
Erlenmeyer pattern
Manufactured from Pyrex™ borosilicate glass
Excellent thermal performance
High resistance to chemical attack
White enamel graduations and marking spot
Graduated to capacity
Printed with Trace Code for downloadable batch certificate
Specifications
Certifications/Compliance
ISO 24450, DIN 12385
Capacity (Metric)
250 mL
Material
Borosilicate Glass, Borosilicate Glass
Graduations
Yes
Height (Metric)
140 mm
Includes
Approximate graduations
Shape
Conical
Writing Surface
Yes
Graduated
Yes
Diameter (Metric) Outer
85 mm
Closure Size
45