HiTrap™ TALON™ Crude Prepacked Columns
HiTrap™ TALON™ Crude Prepacked Columns
Actual product may vary
Fisherbrand
HiTrap™ TALON™ Crude Prepacked Columns
For purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC). Cytiva Lifescience™ HiTrap™ TALON™ Crude Prepacked Columns offer a cobalt-based medium allowing protein purification under native or denaturing conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FKS120341185 x 5 mL
FKS116188935 x 1 mL
Catalog number FKS12034118
Price (SGD)
-
Quantity:
5 x 5 mL
Cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. Allows protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.
  • Prepacked with cobalt-based chromatography media
  • Binding capacity approximately 7mg MBP
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL columns
Media
  • Ligand: TALON™ Superflow Cobalt-IMAC
  • Matrix: 6% cross-linked agarose
  • Average particle size: 60 to 160μm
  • pH stability working range: 3 to 12
1mL columns
  • Column I.D.: 7mm
  • Flow velocity: 1 to 4mL/min.
5mL columns
  • Column I.D.: 16
  • Flow velocity: 5 to 20mL/min.
Specifications
Quantity5 x 5 mL
For Use With (Application)For PurifiCatIon of Histidine-Tagged Recombinant Proteins by Immobilized Metal Affinity Chromatography, Used Buffers and Easily Regenerated with 0.5 M NaOH
Storage Requirements4°C to 8°C
Binding CapacityUp to 20 mg Histidine-tagged Protein1
Diameter (English) Inner0.62 in.
Product TypeHiTrap Talon crude prepacked Column
Diameter (Metric) Inner1.6 cm
Flow Rate1 to 4 mL/min
Particle Size60 to 160 μm
MaterialPolypropylene
Max. Pressure3 bar
Matrix6% Cross-Linked Agarose
pH Range3 to 12
Storage Buffer20% Ethanol
Ligand TypeTalon Superflow Cobalt-IMAC
Packaging Quantity5 x 5 mL
Unit SizePK