Check Order Status
Stuart™ Membrane Filters
Fisherbrand
Stuart™ Membrane Filters
Used with Bibbyjet Pro™ Pipette Controller. Stuart™ Membrane Filters are available in two sizes, and either sterile or non-sterile.
Have Questions?
Catalog Number
Pore Size
Sterility
Quantity
FKS12028681
0.2 μm
Non-sterile
10 Pack
Catalog number
FKS12028681
Price (SGD)
707.00
PK
Add to cart
Pore Size:
0.2 μm
Sterility:
Non-sterile
Quantity:
10 Pack
Price (SGD)
707.00
PK
Add to cart
For use with Bibbyjet Pro™ Pipette Controller:
Lightweight and ergonomic
Powerful enough to fill a 50 mL pipet in just 10 s
Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
Pipet holder fully autoclavable
Venting feature allows air sucked from the pipet to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Specifications
Pore Size
0.2 μm
Sterility
Non-sterile
Type
Filter
For Use With (Equipment)
Bibbyjet Pro Pipette Controller PC2000/2001
Quantity
10 Pack